Assay Details
Target Gene Details
Entrez Gene ID: | 25896 |
Gene Name: | integrator complex subunit 7 |
Gene Aliases: |
C1orf73, INT7 |
Location: |
Chr.1:211940328-212035660 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS7 | NM_001199809.1 | 19 | 3670 | NP_001186738.1 |
| NM_001199811.1 | 20 | 3775 | NP_001186740.1 | |
| NM_001199812.1 | 20 | 3757 | NP_001186741.1 | |
| NM_015434.3 | 20 | 3817 | NP_056249.1 | |
| NR_037667.1 | 21 | 3851 | ||
| XM_011509396.2 | 21 | 3810 | XP_011507698.1 | |
| AL133049.2 | 15 | 2986 | CAB61376.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs187489037] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map