Assay Details
Target Gene Details
Entrez Gene ID: | 1889 |
Gene Name: | endothelin converting enzyme 1 |
Gene Aliases: |
ECE |
Location: |
Chr.1:21217247-21345541 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECE1 | NM_001113348.1 | NP_001106819.1 | ||
| XM_011540872.2 | XP_011539174.1 | |||
| XM_011540873.2 | XP_011539175.1 | |||
| XM_017000511.1 | XP_016856000.1 | |||
| DB290492.1 | ||||
| X98272.1 | CAA66922.1 |
Target Gene Details
Entrez Gene ID: | 100506801 |
Gene Name: | uncharacterized LOC100506801 |
Gene Aliases: |
- |
Location: |
Chr.1:21293290-21299869 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506801 | NR_038404.1 | 3 | 997 |
More Information
Set Membership: |
|

Genomic Map