Assay Details
Target Gene Details
Entrez Gene ID: | 26052 |
Gene Name: | dynamin 3 |
Gene Aliases: |
Dyna III |
Location: |
Chr.1:171841478-172418466 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Gene Details
Entrez Gene ID: | 100874284 |
Gene Name: | DNM3 intronic transcript 1 |
Gene Aliases: |
- |
Location: |
Chr.1:171864192-171864687 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNM3-IT1 | NR_046845.1 | |||
| AA868330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831893 | Chr.1:171833310 - 172015860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map