Assay Details
Target Gene Details
Entrez Gene ID: | 5321 |
Gene Name: | phospholipase A2 group IVA |
Gene Aliases: |
PLA2G4, cPLA2, cPLA2-alpha |
Location: |
Chr.1:186828900-186989896 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G4A | NM_001311193.1 | NP_001298122.1 | ||
| NM_024420.2 | NP_077734.1 | |||
| XM_011509642.2 | XP_011507944.1 | |||
| AK290336.1 | ||||
| AK302938.1 | ||||
| BC114340.2 | AAI14341.1 | |||
| DA451491.1 | ||||
| M68874.1 | AAA60105.1 | |||
| M72393.1 | AAB00789.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762854 | Chr.1:186577969 - 186878442 on Build GRCh38 | Gain |
|
| nsv548406 | Chr.1:186773815 - 186834094 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map