Assay Details
Target Gene Details
Entrez Gene ID: | 55001 |
Gene Name: | tetratricopeptide repeat domain 22 |
Gene Aliases: |
- |
Location: |
Chr.1:54779696-54802379 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC22 | NM_001114108.1 | NP_001107580.1 | ||
| NM_017904.3 | NP_060374.2 | |||
| XM_011541671.2 | XP_011539973.1 | |||
| XM_017001582.1 | XP_016857071.1 | |||
| AK000626.1 | BAA91293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv166n100 | Chr.1:54741809 - 54804866 on Build GRCh38 | Gain |
|
| nsv1000673 | Chr.1:54630765 - 54818045 on Build GRCh38 | Gain |
|
| nsv546266 | Chr.1:54404143 - 54887697 on Build GRCh38 | Gain |
|
| dgv38e212 | Chr.1:54627539 - 54817638 on Build GRCh38 | Gain |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|

Genomic Map