Assay Details
Target Gene Details
Entrez Gene ID: | 347689 |
Gene Name: | SOX2 overlapping transcript |
Gene Aliases: |
NCRNA00043, SOX2OT |
Location: |
Chr.3:181056680-181742228 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX2-OT | NR_075091.1 | |||
| NR_075092.1 | ||||
| DA281835.1 | ||||
| JQ408703.1 | ||||
| JX504683.1 |

Genomic Map