Assay Details
Target Gene Details
Entrez Gene ID: | 6304 |
Gene Name: | SATB homeobox 1 |
Gene Aliases: |
- |
Location: |
Chr.3:18345388-18445588 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SATB1 | NM_001131010.3 | NP_001124482.1 | ||
| NM_001195470.2 | NP_001182399.1 | |||
| NM_001322871.1 | NP_001309800.1 | |||
| NM_001322872.1 | NP_001309801.1 | |||
| NM_001322873.1 | NP_001309802.1 | |||
| NM_001322874.1 | NP_001309803.1 | |||
| NM_001322875.1 | NP_001309804.1 | |||
| NM_001322876.1 | NP_001309805.1 | |||
| NM_002971.5 | NP_002962.1 | |||
| XM_011533988.2 | XP_011532290.1 | |||
| XM_011533989.2 | XP_011532291.1 | |||
| XM_011533990.2 | 10 | 5581 | XP_011532292.1 | |
| AB209761.1 | BAD92998.1 | |||
| AK127242.1 | ||||
| BC001744.1 | AAH01744.1 | |||
| M97287.1 | AAA60304.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589853 | Chr.3:18344240 - 18386958 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map