Assay Details
Target Gene Details
Entrez Gene ID: | 26084 |
Gene Name: | Rho guanine nucleotide exchange factor 26 |
Gene Aliases: |
CSGEF, HMFN1864, SGEF |
Location: |
Chr.3:154121003-154257827 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF26 | NM_001251962.1 | NP_001238891.1 | ||
| AK127282.1 |
Target Gene Details
Entrez Gene ID: | 100507524 |
Gene Name: | ARHGEF26 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:154024401-154121210 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF26-AS1 | NR_037901.1 |
More Information
Set Membership: |
|

Genomic Map