Assay Details
Target Gene Details
Entrez Gene ID: | 28999 |
Gene Name: | Kruppel like factor 15 |
Gene Aliases: |
KKLF |
Location: |
Chr.3:126288121-126357393 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8547n54 | Chr.3:126296824 - 126330591 on Build GRCh38 | Loss |
|
| nsv997409 | Chr.3:126295062 - 126330463 on Build GRCh38 | Loss |
|

Genomic Map