Assay Details
Target Gene Details
Entrez Gene ID: | 3355 |
Gene Name: | 5-hydroxytryptamine receptor 1F |
Gene Aliases: |
5-HT-1F, 5-HT1F, 5HT6, HTR1EL, MR77 |
Location: |
Chr.3:87792767-87994856 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR1F | NM_001322208.1 | NP_001309137.1 | ||
| NM_001322209.1 | NP_001309138.1 | |||
| NM_001322210.1 | NP_001309139.1 | |||
| XM_005264751.3 | XP_005264808.1 | |||
| XM_011533664.2 | XP_011531966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834758 | Chr.3:87818213 - 87965825 on Build GRCh38 | Gain |
|
| esv3596897 | Chr.3:87803460 - 87899387 on Build GRCh38 | Loss |
|
| nsv460757 | Chr.3:87814130 - 87900583 on Build GRCh38 | Loss |
|
| nsv590918 | Chr.3:87274184 - 87861781 on Build GRCh38 | Gain |
|
| nsv834756 | Chr.3:87714643 - 87889029 on Build GRCh38 | Loss |
|
| esv3596900 | Chr.3:87820260 - 87823510 on Build GRCh38 | Loss |
|
| esv3568889 | Chr.3:87805127 - 87900768 on Build GRCh38 | Loss |
|
| esv3596898 | Chr.3:87811292 - 87840474 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map