Assay Details
Target Gene Details
Entrez Gene ID: | 11073 |
Gene Name: | topoisomerase (DNA) II binding protein 1 |
Gene Aliases: |
TOP2BP1 |
Location: |
Chr.3:133600605-133662380 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOPBP1 | NM_007027.3 | NP_008958.2 | ||
| XM_005247076.3 | XP_005247133.1 | |||
| XM_017005636.1 | XP_016861125.1 | |||
| XM_017005637.1 | XP_016861126.1 | |||
| AB019397.1 | BAA34202.1 | |||
| AK026229.1 | ||||
| AK302584.1 | ||||
| BC126209.1 | ||||
| BC151237.1 | ||||
| D87448.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114946023] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map