Assay Details
Target Gene Details
Entrez Gene ID: | 285313 |
Gene Name: | immunoglobulin superfamily member 10 |
Gene Aliases: |
CMF608 |
Location: |
Chr.3:151435051-151619926 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF10 | XM_011512708.2 | 12 | 9806 | XP_011511010.1 |
| XM_011512709.2 | 11 | 9537 | XP_011511011.1 |
Target Gene Details
Entrez Gene ID: | 116931 |
Gene Name: | mediator complex subunit 12 like |
Gene Aliases: |
NOPAR, TNRC11L, TRALP, TRALPUSH |
Location: |
Chr.3:151085665-151436677 on Build GRCh38 |
Assay Gene Location: | Within Exon 49 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED12L | NM_053002.5 | 43 | 9327 | NP_443728.3 |
| XM_011512390.2 | 44 | 9599 | XP_011510692.1 | |
| XM_017005676.1 | 46 | 9755 | XP_016861165.1 | |
| XM_017005677.1 | 46 | 9752 | XP_016861166.1 | |
| XM_017005678.1 | 44 | 9614 | XP_016861167.1 | |
| XM_017005679.1 | 45 | 9485 | XP_016861168.1 | |
| XM_017005680.1 | 44 | 9473 | XP_016861169.1 | |
| AK124913.1 | 1 | 816 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829757 | Chr.3:151327092 - 151508654 on Build GRCh38 | Loss |
|
| nsv4063 | Chr.3:151408925 - 151444402 on Build GRCh38 | Insertion |
|
| nsv508960 | Chr.3:151425271 - 151443978 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map