Assay Details
Target Gene Details
Entrez Gene ID: | 23228 |
Gene Name: | phospholipase C like 2 |
Gene Aliases: |
PLCE2 |
Location: |
Chr.3:16802691-17090606 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCL2 | XM_017006023.1 | 1 | 91 | XP_016861512.1 |
| AK309946.1 | 1 | 131 |
More Information
Set Membership: |
|

Genomic Map