Assay Details
Target Gene Details
Entrez Gene ID: | 7018 |
Gene Name: | transferrin |
Gene Aliases: |
HEL-S-71p, PRO1557, PRO2086, TFQTL1 |
Location: |
Chr.3:133661998-133779006 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TF | NM_001063.3 | NP_001054.1 | ||
| XM_017007089.1 | XP_016862578.1 | |||
| XM_017007090.1 | XP_016862579.1 | |||
| AJ252279.1 | CAB96907.1 | |||
| AK222755.1 | BAD96475.1 | |||
| AK293842.1 | ||||
| AK295334.1 | ||||
| AK295419.1 | ||||
| AK303753.1 | ||||
| BC059367.1 | AAH59367.1 | |||
| BX648533.1 | ||||
| CR936810.1 | ||||
| DC352309.1 | ||||
| DQ923758.1 | ||||
| GQ472199.1 | ||||
| M12530.1 | AAA61140.1 | |||
| S95936.1 | AAB22049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829729 | Chr.3:133726027 - 133901543 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map