Assay Details
Target Gene Details
Entrez Gene ID: | 5290 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha |
Gene Aliases: |
CLOVE, CWS5, MCAP, MCM, MCMTC, PI3K, PI3K-alpha, p110-alpha |
Location: |
Chr.3:179148114-179240093 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CA | NM_006218.3 | NP_006209.2 | ||
| XM_006713658.3 | XP_006713721.1 | |||
| XM_011512894.2 | XP_011511196.1 | |||
| AK292940.1 | ||||
| BX484734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010571 | Chr.3:179159177 - 179170365 on Build GRCh38 | Gain |
|
| nsv1161006 | Chr.3:179154246 - 179174436 on Build GRCh38 | Duplication |
|
| esv275373 | Chr.3:179169430 - 179174548 on Build GRCh38 | Gain+Loss |
|
| esv2763744 | Chr.3:179165384 - 179170377 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map