Assay Details
Target Gene Details
Entrez Gene ID: | 51409 |
Gene Name: | HemK methyltransferase family member 1 |
Gene Aliases: |
HEMK, MTQ1 |
Location: |
Chr.3:50569152-50589050 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEMK1 | NM_001317851.1 | NP_001304780.1 | ||
| NM_016173.4 | NP_057257.1 | |||
| XM_005265218.3 | XP_005265275.1 | |||
| XM_011533806.2 | XP_011532108.1 | |||
| XM_011533807.1 | XP_011532109.1 | |||
| XM_011533808.1 | XP_011532110.1 | |||
| XM_011533809.2 | XP_011532111.1 | |||
| XM_011533810.2 | XP_011532112.1 | |||
| XM_011533812.2 | XP_011532114.1 | |||
| XM_011533813.2 | XP_011532115.1 | |||
| XM_011533814.2 | XP_011532116.1 | |||
| AF131220.1 | AAD26417.1 | |||
| AF172244.1 | AAD51328.1 | |||
| AK025973.1 | ||||
| AK124369.1 | ||||
| AK314023.1 | ||||
| BC000781.2 | AAH00781.1 | |||
| BX537562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584502 | Chr.3:50327055 - 50622310 on Build GRCh38 | Gain |
|
| nsv590273 | Chr.3:50536782 - 50640720 on Build GRCh38 | Loss |
|
| nsv460532 | Chr.3:50498842 - 50578618 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map