Assay Details
Target Gene Details
Entrez Gene ID: | 9901 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 3 |
Gene Aliases: |
ARHGAP14, MEGAP, SRGAP2, WRP |
Location: |
Chr.3:8980591-9362979 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRGAP3 | NM_001033117.2 | NP_001028289.1 | ||
| NM_014850.3 | NP_055665.1 | |||
| XM_011534295.1 | XP_011532597.1 | |||
| XM_011534296.1 | XP_011532598.1 | |||
| XM_017007576.1 | XP_016863065.1 | |||
| XM_017007577.1 | XP_016863066.1 | |||
| XM_017007580.1 | XP_016863069.1 | |||
| AB007871.2 | BAA24841.2 | |||
| AF427144.1 | AAN07095.1 | |||
| AF464189.1 | AAN57784.1 | |||
| AK094355.1 |
Target Gene Details
Entrez Gene ID: | 100288831 |
Gene Name: | SRGAP3 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.3:9216895-9219508 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRGAP3-AS3 | NR_103443.1 | |||
| AK302139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3690 | Chr.3:9192267 - 9223982 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map