Assay Details
Target Gene Details
Entrez Gene ID: | 100862662 |
Gene Name: | ALDH1L1 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.3:126180065-126210169 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH1L1-AS2 | NR_046383.1 | |||
| DA256650.1 | ||||
| DA566603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8545n54 | Chr.3:126091128 - 126251891 on Build GRCh38 | Loss |
|
| nsv3985 | Chr.3:126173880 - 126218546 on Build GRCh38 | Deletion |
|
| nsv1007256 | Chr.3:126163635 - 126238737 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76222957] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map