Assay Details
Target Gene Details
Entrez Gene ID: | 2257 |
Gene Name: | fibroblast growth factor 12 |
Gene Aliases: |
FGF12B, FHF1 |
Location: |
Chr.3:192139393-192727599 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF12 | NM_004113.5 | 6 | 4183 | NP_004104.3 |
| NM_021032.4 | 5 | 4949 | NP_066360.1 | |
| XM_005247227.2 | 5 | 4147 | XP_005247284.1 | |
| XM_006713538.3 | 5 | 4015 | XP_006713601.1 | |
| XM_017005879.1 | 5 | 4034 | XP_016861368.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5007n100 | Chr.3:191641572 - 192217259 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map