Assay Details
Target Gene Details
Entrez Gene ID: | 22871 |
Gene Name: | neuroligin 1 |
Gene Aliases: |
NL1 |
Location: |
Chr.3:173395899-174291287 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLGN1 | XM_011512552.2 | XP_011510854.1 | ||
| XM_017005892.1 | XP_016861381.1 | |||
| XM_017005893.1 | XP_016861382.1 | |||
| XM_017005896.1 | XP_016861385.1 | |||
| XM_017005899.1 | XP_016861388.1 | |||
| XM_017005901.1 | XP_016861390.1 | |||
| XM_017005904.1 | XP_016861393.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73046079] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map