Assay Details
Target Gene Details
Entrez Gene ID: | 6259 |
Gene Name: | receptor-like tyrosine kinase |
Gene Aliases: |
D3S3195, JTK5, JTK5A, RYK1 |
Location: |
Chr.3:134157133-134250834 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RYK | NM_001005861.2 | NP_001005861.1 | ||
| NM_002958.3 | NP_002949.2 | |||
| XM_017007016.1 | XP_016862505.1 | |||
| BC021700.2 | AAH21700.2 | |||
| S59184.1 | AAB26341.1 | |||
| X69970.1 | CAA49591.1 | |||
| X96588.1 | CAA65406.1 |
More Information
Set Membership: |
|

Genomic Map