Assay Details
Target Gene Details
Entrez Gene ID: | 3680 |
Gene Name: | integrin subunit alpha 9 |
Gene Aliases: |
ALPHA-RLC, ITGA4L, RLC |
Location: |
Chr.3:37452322-37819790 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA9 | NM_002207.2 | NP_002198.2 | ||
| BC030198.1 | AAH30198.1 | |||
| D25303.1 | BAA04984.1 | |||
| L24158.1 | AAA16099.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3595886 | Chr.3:37316808 - 37528893 on Build GRCh38 | Gain |
|
| esv3595885 | Chr.3:37316562 - 37455864 on Build GRCh38 | Gain |
|
| esv3595887 | Chr.3:37319289 - 37878005 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78665795] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map