Assay Details
Target Gene Details
Entrez Gene ID: | 23228 |
Gene Name: | phospholipase C like 2 |
Gene Aliases: |
PLCE2 |
Location: |
Chr.3:16802691-17090606 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLCL2 | XM_017006023.1 | XP_016861512.1 | ||
| AK309946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv460454 | Chr.3:16809700 - 16832586 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map