Assay Details
Target Gene Details
Entrez Gene ID: | 339926 |
Gene Name: | EHHADH antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:185162901-185191955 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EHHADH-AS1 | NR_038990.1 | |||
| AL703608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5002n100 | Chr.3:185129229 - 185287036 on Build GRCh38 | Gain |
|

Genomic Map