Assay Details
Target Gene Details
Entrez Gene ID: | 4292 |
Gene Name: | mutL homolog 1 |
Gene Aliases: |
COCA2, FCC2, HNPCC, HNPCC2, hMLH1 |
Location: |
Chr.3:36993350-37050846 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834662 | Chr.3:36830593 - 37033470 on Build GRCh38 | Loss |
|
| esv3568690 | Chr.3:37022671 - 37029347 on Build GRCh38 | Loss |
|
| nsv834663 | Chr.3:36966258 - 37148914 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map