Assay Details
Target Gene Details
Entrez Gene ID: | 8419 |
Gene Name: | beaded filament structural protein 2 |
Gene Aliases: |
CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN |
Location: |
Chr.3:133397882-133475212 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BFSP2 | NM_003571.3 | NP_003562.1 | ||
| XM_017007315.1 | XP_016862804.1 | |||
| BC113518.1 | ||||
| BC113520.1 | ||||
| BM696550.1 | ||||
| U48224.1 | AAC50414.1 |
Target Gene Details
Entrez Gene ID: | 85003 |
Gene Name: | BFSP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:133426402-133491146 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BFSP2-AS1 | NR_135276.1 | |||
| NR_135277.1 | ||||
| NR_135278.1 | ||||
| AA746278.1 | ||||
| HY096628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4903n100 | Chr.3:133418820 - 133467964 on Build GRCh38 | Loss |
|
| nsv522281 | Chr.3:133418590 - 133448519 on Build GRCh38 | Gain |
|
| nsv819448 | Chr.3:133426609 - 133446741 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115023461] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map