Assay Details
Target Gene Details
Entrez Gene ID: | 22871 |
Gene Name: | neuroligin 1 |
Gene Aliases: |
NL1 |
Location: |
Chr.3:173395899-174291287 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Gene Details
Entrez Gene ID: | 100874010 |
Gene Name: | NLGN1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:173910498-173920796 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLGN1-AS1 | NR_046664.1 | |||
| BX101530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003504 | Chr.3:173883091 - 174247789 on Build GRCh38 | Gain |
|
| esv3575641 | Chr.3:173912877 - 173986041 on Build GRCh38 | Gain |
|
| dgv4987n100 | Chr.3:173907550 - 173993420 on Build GRCh38 | Gain |
|
| dgv305n21 | Chr.3:173856727 - 174253013 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map