Assay Details
Target Gene Details
Entrez Gene ID: | 10425 |
Gene Name: | ariadne RBR E3 ubiquitin protein ligase 2 |
Gene Aliases: |
ARI2, TRIAD1 |
Location: |
Chr.3:48918820-48985541 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
More Information
Set Membership: |
|

Genomic Map