Assay Details
Target Gene Details
Entrez Gene ID: | 25924 |
Gene Name: | myosin VIIA and Rab interacting protein |
Gene Aliases: |
SLAC2-C, SLAC2C |
Location: |
Chr.3:39808914-40260321 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYRIP | NM_001284423.1 | NP_001271352.1 | ||
| NM_001284424.1 | NP_001271353.1 | |||
| NM_001284425.1 | NP_001271354.1 | |||
| NM_015460.3 | NP_056275.2 | |||
| NR_104316.1 | ||||
| XM_011533575.1 | XP_011531877.1 | |||
| AF396687.1 | AAM43954.1 | |||
| AK126013.1 | ||||
| AK310972.1 | ||||
| AK312412.1 | ||||
| BC092512.1 | AAH92512.1 | |||
| BC109311.1 | AAI09312.1 | |||
| BC109312.1 | AAI09313.1 | |||
| CF138044.1 | ||||
| DN999201.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523927 | Chr.3:39867354 - 39890024 on Build GRCh38 | Loss |
|
| nsv3773 | Chr.3:39868257 - 39901670 on Build GRCh38 | Insertion |
|
| nsv517803 | Chr.3:39859533 - 39991467 on Build GRCh38 | Gain |
|
| esv3595939 | Chr.3:39872952 - 39884999 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map