Assay Details
Target Gene Details
Entrez Gene ID: | 285315 |
Gene Name: | chromosome 3 open reading frame 33 |
Gene Aliases: |
AC3-33 |
Location: |
Chr.3:155762612-155806287 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf33 | NM_001308229.1 | NP_001295158.1 | ||
| NM_173657.2 | NP_775928.1 | |||
| XM_005247379.4 | XP_005247436.1 | |||
| XM_011512710.2 | XP_011511012.1 | |||
| XM_011512711.2 | XP_011511013.1 | |||
| AF115515.1 | AAO06952.1 | |||
| AK055701.1 | BAB70989.1 | |||
| AK289890.1 | ||||
| AL832165.1 | ||||
| BC042198.1 | AAH42198.1 | |||
| BC064898.1 | AAH64898.1 | |||
| DA275323.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4928n100 | Chr.3:155763308 - 155816827 on Build GRCh38 | Gain |
|
| esv3598283 | Chr.3:155759067 - 155869415 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map