Assay Details
Target Gene Details
Entrez Gene ID: | 80014 |
Gene Name: | WW and C2 domain containing 2 |
Gene Aliases: |
BOMB |
Location: |
Chr.4:183098677-183320777 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWC2 | NM_024949.5 | NP_079225.5 | ||
| XM_011532269.2 | XP_011530571.1 | |||
| DN999977.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016376 | Chr.4:182859067 - 183262599 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map