Assay Details
Target Gene Details
Entrez Gene ID: | 80014 |
Gene Name: | WW and C2 domain containing 2 |
Gene Aliases: |
BOMB |
Location: |
Chr.4:183098677-183320777 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWC2 | NM_024949.5 | NP_079225.5 | ||
| XM_011532269.2 | XP_011530571.1 | |||
| AK126057.1 | BAC86418.1 | |||
| AK127061.1 | BAC86807.1 | |||
| BM046889.1 | ||||
| BX647378.1 | CAH10569.1 |
Target Gene Details
Entrez Gene ID: | 101928734 |
Gene Name: | WWC2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.4:183233628-183240634 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWC2-AS1 | NR_126473.1 | |||
| BX537950.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1016376 | Chr.4:182859067 - 183262599 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map