Assay Details
Target Gene Details
Entrez Gene ID: | 3295 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 4 |
Gene Aliases: |
DBP, MFE-2, MPF-2, PRLTS1, SDR8C1 |
Location: |
Chr.5:119452443-119542335 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSD17B4 | NM_000414.3 | NP_000405.1 | ||
| NM_001199291.2 | NP_001186220.1 | |||
| NM_001199292.1 | NP_001186221.1 | |||
| NM_001292027.1 | NP_001278956.1 | |||
| NM_001292028.1 | NP_001278957.1 | |||
| AK094049.1 | ||||
| AK293254.1 | ||||
| AK298075.1 | ||||
| AK299685.1 | ||||
| AK301212.1 | ||||
| AK312449.1 | ||||
| BC003098.1 | AAH03098.1 | |||
| DA296575.1 | ||||
| DA426062.1 | ||||
| DA508630.1 | ||||
| DC325873.1 | ||||
| KU178135.1 | ||||
| KU178136.1 | ||||
| KU178137.1 | ||||
| KU178138.1 | ||||
| X87176.1 | CAA60643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4974 | Chr.5:119414564 - 119459681 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map