Assay Details
Target Gene Details
Entrez Gene ID: | 101927078 |
Gene Name: | uncharacterized LOC101927078 |
Gene Aliases: |
- |
Location: |
Chr.5:114447418-114773413 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927078 | NR_130785.1 | |||
| AK097686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830452 | Chr.5:114462920 - 114639531 on Build GRCh38 | Loss |
|
| esv3606342 | Chr.5:114501210 - 114535659 on Build GRCh38 | Loss |
|
| nsv599433 | Chr.5:114491590 - 114565393 on Build GRCh38 | Gain |
|

Genomic Map