Assay Details
Target Gene Details
Entrez Gene ID: | 100508120 |
Gene Name: | GMDS antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.6:2245753-2413591 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMDS-AS1 | NR_046229.1 | |||
| AK023629.1 | 5 | 1554 | ||
| CN395927.1 | ||||
| CR739457.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607900 | Chr.6:2093592 - 2775672 on Build GRCh38 | Gain |
|
| esv2763526 | Chr.6:2229830 - 2311682 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73342705] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map