Assay Details
Target Gene Details
Entrez Gene ID: | 100507462 |
Gene Name: | GVQW motif containing 2 |
Gene Aliases: |
- |
Location: |
Chr.6:138725211-138773679 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GVQW2 | NM_001242740.1 | NP_001229669.1 | ||
| AK092592.1 | ||||
| DN917422.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3610962 | Chr.6:138741041 - 138754403 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73572712] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map