Assay Details
Target Gene Details
Entrez Gene ID: | 7172 |
Gene Name: | thiopurine S-methyltransferase |
Gene Aliases: |
TPMTD |
Location: |
Chr.6:18128311-18155165 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPMT | NM_000367.3 | NP_000358.1 | ||
| XM_011514839.2 | XP_011513141.1 | |||
| XM_017011241.1 | XP_016866730.1 | |||
| AF021876.1 | AAB80746.1 | |||
| AF021877.1 | AAB80747.1 | |||
| AK310420.1 | ||||
| BC005339.1 | AAH05339.1 | |||
| BC009596.1 | AAH09596.1 | |||
| BI598325.1 | ||||
| DB481604.1 | ||||
| S62904.1 | AAB27277.1 | |||
| U12387.1 | AAC50130.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034755 | Chr.6:18064320 - 18173781 on Build GRCh38 | Loss |
|
| nsv601050 | Chr.6:18134947 - 18221240 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map