Assay Details
Target Gene Details
Entrez Gene ID: | 57531 |
Gene Name: | HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
SPPRS |
Location: |
Chr.6:104682670-104859919 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HACE1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1024939 | Chr.6:104608417 - 104767557 on Build GRCh38 | Loss |
|

Genomic Map