Assay Details
Target Gene Details
Entrez Gene ID: | 100507462 |
Gene Name: | GVQW motif containing 2 |
Gene Aliases: |
- |
Location: |
Chr.6:138725211-138773679 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GVQW2 | NM_001242740.1 | NP_001229669.1 | ||
| AK092592.1 | ||||
| DN917422.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114394034,rs116779877] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map