Assay Details
Target Gene Details
Entrez Gene ID: | 23118 |
Gene Name: | TGF-beta activated kinase 1/MAP3K7 binding protein 2 |
Gene Aliases: |
CHTD2, MAP3K7IP2, TAB-2 |
Location: |
Chr.6:149217924-149411613 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAB2 | NM_001292035.2 | NP_001278964.1 | ||
| AK295750.1 | ||||
| BX645362.1 | ||||
| DA154923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5531 | Chr.6:149237991 - 149253458 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map