Assay Details
Target Gene Details
Entrez Gene ID: | 55510 |
Gene Name: | DEAD-box helicase 43 |
Gene Aliases: |
CT13, HAGE |
Location: |
Chr.6:73394562-73417566 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX43 | NM_018665.2 | NP_061135.2 | ||
| XM_011535926.1 | XP_011534228.1 | |||
| XM_011535927.2 | XP_011534229.1 | |||
| XM_011535928.2 | XP_011534230.1 | |||
| XM_017011003.1 | XP_016866492.1 | |||
| AJ278110.1 | CAB92442.1 | |||
| AK303324.1 | ||||
| AK313686.1 | ||||
| AL136751.1 | CAB66685.1 | |||
| BC066938.1 | AAH66938.1 | |||
| BX646425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830693 | Chr.6:73339619 - 73506357 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map