Assay Details
Target Gene Details
Entrez Gene ID: | 5238 |
Gene Name: | phosphoglucomutase 3 |
Gene Aliases: |
AGM1, IMD23, PAGM, PGM 3 |
Location: |
Chr.6:83150728-83193936 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGM3 | NM_001199917.1 | NP_001186846.1 | ||
| XM_017010936.1 | XP_016866425.1 | |||
| AK301867.1 | ||||
| DB033430.1 |
Target Gene Details
Entrez Gene ID: | 112611 |
Gene Name: | RWD domain containing 2A |
Gene Aliases: |
RWDD2, dJ747H23.2 |
Location: |
Chr.6:83193313-83198935 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RWDD2A | NM_001322335.1 | NP_001309264.1 | ||
| NM_001322336.1 | NP_001309265.1 | |||
| NM_001322337.1 | NP_001309266.1 | |||
| NM_001322339.1 | NP_001309268.1 | |||
| NM_033411.4 | NP_219479.2 | |||
| XM_017010222.1 | XP_016865711.1 | |||
| XM_017010223.1 | XP_016865712.1 | |||
| AK295724.1 | ||||
| AK302978.1 | ||||
| AK309249.1 | ||||
| AK313083.1 | ||||
| BG676907.1 | ||||
| CD108545.1 | ||||
| DC377982.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map