Assay Details
Target Gene Details
Entrez Gene ID: | 222611 |
Gene Name: | adhesion G protein-coupled receptor F2 |
Gene Aliases: |
GPR111, PGR20, hGPCR35 |
Location: |
Chr.6:47656590-47697797 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRF2 | NM_153839.6 | NP_722581.4 | ||
| AY140953.1 | AAN46667.1 | |||
| DQ315369.1 | ABC41928.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26618 | Chr.6:47654534 - 47685135 on Build GRCh38 | Gain |
|
| nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map