Assay Details
Target Gene Details
Entrez Gene ID: | 55362 |
Gene Name: | transmembrane protein 63B |
Gene Aliases: |
C6orf110 |
Location: |
Chr.6:44126628-44155519 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM63B | NM_001318792.1 | NP_001305721.1 | ||
| NM_018426.2 | NP_060896.1 | |||
| XM_005249211.3 | XP_005249268.1 | |||
| XM_005249213.4 | XP_005249270.1 | |||
| XM_005249215.4 | XP_005249272.1 | |||
| XM_005249217.1 | 1 | 64 | XP_005249274.1 | |
| XM_006715135.1 | XP_006715198.1 | |||
| XM_017010999.1 | XP_016866488.1 | |||
| XM_017011000.1 | XP_016866489.1 | |||
| XM_017011001.1 | XP_016866490.1 | |||
| XM_017011002.1 | 1 | 72 | XP_016866491.1 | |
| AK093629.1 | BAC04207.1 | |||
| AK309791.1 | ||||
| BE463633.1 | ||||
| CR627460.1 | CAH10540.1 | |||
| DC408120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1073976 | Chr.6:44127062 - 44127763 on Build GRCh38 | Deletion |
|
| nsv603002 | Chr.6:44078406 - 44149870 on Build GRCh38 | Loss |
|
| nsv509132 | Chr.6:44123721 - 44185060 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map