Assay Details
Target Gene Details
Entrez Gene ID: | 64065 |
Gene Name: | PERP, TP53 apoptosis effector |
Gene Aliases: |
KCP1, KRTCAP1, PIGPC1, THW, dJ496H19.1 |
Location: |
Chr.6:138088505-138107523 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PERP | NM_022121.4 | NP_071404.2 | ||
| AF317550.1 | AAG35063.1 | |||
| AJ251830.1 | CAC17766.1 | |||
| AK074585.1 | BAC11074.1 | |||
| AK075082.1 | BAC11390.1 | |||
| AK093516.1 | ||||
| AK097387.1 | ||||
| AK097958.1 | BAC05205.1 | |||
| AK225907.1 | ||||
| AK314526.1 | ||||
| AY157578.1 | AAO13162.1 | |||
| BC010163.2 | AAH10163.1 | |||
| CX784592.1 | ||||
| DA750046.1 |
More Information
Set Membership: |
|

Genomic Map