Assay Details
Target Gene Details
Entrez Gene ID: | 346157 |
Gene Name: | zinc finger protein 391 |
Gene Aliases: |
dJ153G14.3 |
Location: |
Chr.6:27374635-27403908 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF391 | NM_001076781.2 | NP_001070249.1 | ||
| NM_001322288.1 | NP_001309217.1 | |||
| NM_001322289.1 | NP_001309218.1 | |||
| NM_001322293.1 | NP_001309222.1 | |||
| XM_005249082.3 | XP_005249139.1 | |||
| XM_006715086.2 | XP_006715149.1 | |||
| XM_011514569.1 | XP_011512871.1 | |||
| AK294965.1 | ||||
| BG722372.1 | ||||
| DA758693.1 | ||||
| DB221901.1 | ||||
| DB505213.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5234 | Chr.6:27377838 - 27410246 on Build GRCh38 | Insertion |
|
| nsv5235 | Chr.6:27390719 - 27436295 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map