Assay Details
Target Gene Details
Entrez Gene ID: | 2898 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 2 |
Gene Aliases: |
EAA4, GLR6, GLUK6, GLUR6, GluK2, MRT6 |
Location: |
Chr.6:101393708-102070083 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIK2 | XM_005266945.2 | XP_005267002.1 | ||
| HM149340.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs571279489] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map