Assay Details
Target Gene Details
Entrez Gene ID: | 55788 |
Gene Name: | LMBR1 domain containing 1 |
Gene Aliases: |
C6orf209, LMBD1, MAHCF, NESI |
Location: |
Chr.6:69675749-69797157 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMBRD1 | NM_018368.3 | NP_060838.3 | ||
| XM_011535941.1 | XP_011534243.1 | |||
| AF113224.1 | AAG39295.1 | |||
| AF208863.1 | AAF64277.1 | |||
| AF211480.1 | AAK26247.1 | |||
| AK002102.1 | BAA92087.1 | |||
| AK021916.1 | 1 | 436 | ||
| AK225187.1 | ||||
| AK225537.1 | ||||
| AK290069.1 | ||||
| AY136817.1 | AAN11301.1 | |||
| BC010360.1 | AAH10360.1 | |||
| BC047073.1 | AAH47073.1 | |||
| BX647737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830682 | Chr.6:69687959 - 69882991 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map