Assay Details
Target Gene Details
Entrez Gene ID: | 3096 |
Gene Name: | human immunodeficiency virus type I enhancer binding protein 1 |
Gene Aliases: |
CIRIP, CRYBP1, GAAP, MBP-1, PRDII-BF1, Schnurri-1, ZAS1, ZNF40, ZNF40A |
Location: |
Chr.6:12002096-12212034 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIVEP1 | XM_011514546.2 | 2 | 419 | XP_011512848.1 |
| XM_011514548.2 | XP_011512850.1 | |||
| XM_011514551.2 | 2 | 418 | XP_011512853.1 | |
| XM_017010800.1 | 3 | 305 | XP_016866289.1 | |
| XM_017010801.1 | 3 | 246 | XP_016866290.1 | |
| XM_017010802.1 | 1 | 325 | XP_016866291.1 | |
| XM_017010803.1 | 2 | 420 | XP_016866292.1 | |
| XM_017010804.1 | 2 | 423 | XP_016866293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv22880 | Chr.6:12007827 - 12013566 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map