Assay Details
Target Gene Details
Entrez Gene ID: | 102724422 |
Gene Name: | uncharacterized LOC102724422 |
Gene Aliases: |
- |
Location: |
Chr.6:100880867-100896666 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724422 | XM_017011598.1 | XP_016867087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3610149 | Chr.6:100750184 - 100963770 on Build GRCh38 | Gain |
|
| esv2763587 | Chr.6:100829998 - 101062615 on Build GRCh38 | Loss |
|
| esv3571321 | Chr.6:100875185 - 100895907 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113052923] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map